Learn More
Tocris Bioscience™ VH 298
High-affinity inhibitor of VHL
Brand: Tocris Bioscience™ 6156/10
Description
VH 298 is a high-affinity inhibitor of E3 ubiquitin ligase VHL (Kd = 80-90 nM). Blocks interaction between VHL and HIF-α downstream of HIF-α hydroxylation, initiating hypoxic response. Results in time- and concentration-dependent accumulation of hydroxylated HIF-α, and upregulates mRNA and protein levels of HIF target genes, with similar transcriptional profile to hypoxia. Cell permeable.Negative control cis VH 298 also available.

Specifications
Inhibitor | |
C27H33N5O4S | |
Store at -20°C | |
10 mg |
Ubiquitin Ligase (E3) | |
523.65g/mol | |
Ubiquitin Ligase (E3) Inhibitors |
Product Suggestions
Customers who viewed this item also viewed
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only