missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ VH 298

High-affinity inhibitor of VHL

£239.00 - £917.00

Specifications

Product Type Inhibitor
Target Proteins Ubiquitin Ligase (E3)
Molecular Formula C27H33N5O4S
Formula Weight 523.65g/mol
Content And Storage Store at -20°C
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18721378
View Documents
Tocris Bioscience™
6156/10
10 mg
£239.00
10mg
Estimated Shipment: 10-04-2025
to see stock.
Add to basket
 
18770268
View Documents
Tocris Bioscience™
6156/50
50 mg
£917.00
50mg
Estimated Shipment: 10-04-2025
to see stock.
Add to basket
 
Description

Description

High-affinity inhibitor of E3 ubiquitin ligase VHL (Kd = 80-90 nM). Blocks interaction between VHL and HIF-α downstream of HIF-α hydroxylation, initiating hypoxic response. Results in time- and concentration-dependent accumulation of hydroxylated HIF-α, and upregulates mRNA and protein levels of HIF target genes, with similar transcriptional profile to hypoxia. Cell permeable.

Negative control cis VH 298 also available.

Specifications

Specifications

Inhibitor
C27H33N5O4S
Store at -20°C
Ubiquitin Ligase (E3)
523.65g/mol
Ubiquitin Ligase (E3) Inhibitors
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Tocris Bioscience™ VH 298

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.