Learn More
Invitrogen™ NBD C6-Ceramide Complexed to BSA
Selective stain for Golgi apparatus in live and fixed cells
Brand: Invitrogen™ N22651
266.01 GBP valid until 2025-03-21
Use promo code "24111" to get your promotional price.
303.24 GBP valid until 2025-04-18
Use promo code "24107" to get your promotional price.
Description
NBD C6-ceramide can be used to study sphingolipid transport and metabolism mechanisms. Complexing fluorescent lipids with bovine serum albumin (BSA) facilitates cell labeling by eliminating the need for organic solvents to dissolve the lipophilic probe - the BSA-complexed probe can be directly dissolved in water. The environment sensitive NBD dye is weakly fluorescent in water but increases in aprotic solvents and other nonpolar environments with excitation/emission maxima ∽466/536nm.
Cell Analysis, Cell Structure, Golgi
Order Info
Shipping Condition: Room Temperature

Specifications
Sphingolipids | |
5 mg | |
Solid | |
Invitrogen |
Product Suggestions
Customers who viewed this item also viewed.
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only. Not for use in diagnostic procedures.