Learn More
Invitrogen™ PFB-FDGlu (5-(Pentafluorobenzoylamino)Fluorescein Di-β-D-Glucopyranoside)
PFB-FDGlu is a cell-permeable substrate used for live cell imaging of GCase activity. It fluoresces green upon cleavage by lysosomal GCase, facilitating specific, single-cell analysis via high-content imaging, flow cytometry, and microscopy.
Brand: Invitrogen™ P11947
439.28 GBP valid until 2025-04-18
Use promo code "24107" to get your promotional price.
Description
PFB-FDGlu (5-(Pentafluorobenzoylamino) Fluorescein Di-beta-D-Glucopyranoside) is a glycosidase activity probe for live cell imaging of lysosomal glucocerebrosidase (GCase) activity. This enzyme is vital for glycolipid metabolism, and its deficiency is linked to diseases like Gaucher’s and Parkinson's. PFB-FDGlu fluoresces green upon cleavage by GCase, allowing specific, single-cell analysis via flow cytometry, fluorescent microscopy, and high-content imaging.
PFB-FDGlu is cell-permeable and when loaded into cells, localizing to endosomal and lysosomal compartments while maintaining cell viability. Upon enzymatic cleavage by active GCase enzymes, it yields a green-fluorescent dye (PFB-F) with excitation/emission maxima of approximately 492/516 nm. This fluorescence can be detected using fluorescence microscopy, high-content imaging, and flow cytometry, allowing for specific, single-cell analysis of GCase activity.
Advantages of PFB-FDGlu for detecting GCase activity in lysosomes:
- High specificity—utilizes a lysosomal GCase substrate for measurement of GCase activity, ensuring specific lysosomal staining (note that with long incubations the fluorescent PFB-F will migrate to the cytoplasm)
- Live cell imaging—allows for the assessment of GCase activity in intact, living cells with minimal impact on cell viability, even with extended incubation
- Single-cell analysis—enables specific, single-cell analysis with flow cytometry, high-content imaging, and microscopy to distinguish between different cell types
- Bright fluorescence—yields a bright green fluorescent dye upon cleavage by GCase, enhancing detection sensitivity
- Versatile protocol—can be combined with other phenotypic indicators, such as viability stains and antibodies, to identify distinct cell populations or as a selection criterion for cell sorting
GCase, encoded by the GBA1 gene, is a lysosomal hydrolase that converts glucosylceramide to glucose and ceramide. Mutations in GBA1 leading to GCase deficiency can cause lysosomal storage disorders. GCase deficiencies are implicated in several diseases, including Gaucher's disease and Parkinson's disease.
Order Info
Shipping Condition: Room Temperature

Specifications
Cell-Permeant | |
Store in freezer (-5°C to -30°C). | |
Powder | |
GCase Substrate | |
Beta-Glucosidase Substrate | |
492/526 | |
Fluorescein | |
Room Temperature |
Green | |
Fluorescence | |
5 mg | |
Chemical Substrate | |
Beta-Glucosidase | |
Flow Cytometer, High Content Imaging Instrument, Fluorescent Microscope, Confocal Microscope | |
Substrate | |
Freezer (-5°C to -30°C) |
Product Suggestions
Customers who viewed this item also viewed
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Special Offers
-
Multiple Manufacturers
Powering Proteomics
Advance your proteomics research with essential toolsPromo Code: 25977Expires: 2025-05-31
Days
54Hours
19Minutes
12Seconds
59New Offer Save Up to 92%
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only. Not for use in diagnostic procedures.