missing translation for 'onlineSavingsMsg'
Learn More
Molybdenum(IV) sulfide, 98%, Thermo Scientific Chemicals Product Code.: 11386618

Molybdenum(IV) sulfide, 98%, Thermo Scientific Chemicals

Product Code. 11386618
250 g
Click to view available options
Quantity:
250 g
1 kg
This item is not returnable. View return policy

Product Code. 11386618

Brand: Thermo Scientific Alfa Aesar 012213.30

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Hydrogenation and Isomerization Catalyst, Dry Lubricant

It is widely used as a solid lubricant .It finds applications mainly as Intermediates, Process regulators, Processing aids. Employed as a co catalyst for desulfurization in petrochemistry. As a catalyst in the electrolysis of water.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1317-33-5
Molecular Formula MoS2
Molecular Weight (g/mol) 160.07
MDL Number MFCD00003470
InChI Key CWQXQMHSOZUFJS-UHFFFAOYSA-N
Synonym molybdenum disulfide, molybdenite, molybdenum iv sulfide, molybdenum disulphide, molysulfide, molykote, motimol, nichimoly c, sumipowder pa, molykote zShow MoreShow Less
PubChem CID 14823
ChEBI CHEBI:30704
SMILES S=[Mo]=S

Specifications

Melting Point 1185°C
Color Black
Physical Form Powder
Quantity 250 g
Assay Percent Range 98%
Solubility Information Insoluble in water and dilute acids
IUPAC Name disulfanylidenemolybdenum
Formula Weight 160.08
Odor Odorless
Appearance Black powder
Chemical Name or Material Molybdenum(IV) sulfide
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberT24K009 Certificate TypeCertificate of Analysis Date30/07/2024 Catalog Number012213.A1,012213.30
Lot NumberT12J001 Certificate TypeCertificate of Analysis Date19/07/2023 Catalog Number012213,012213.A1,012213.30
Lot NumberP02J022 Certificate TypeCertificate of Analysis Date09/05/2023 Catalog Number012213.30
Lot NumberS28I047 Certificate TypeCertificate of Analysis Date11/07/2022 Catalog Number012213.A1,012213.30
Lot NumberT09E025 Certificate TypeCertificate of Analysis Date12/04/2022 Catalog Number012213.A1,012213.30
compliance-icons
Product Identifier
  • Molybdenum(IV) sulfide
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H332-Harmful if inhaled.
Precautionary Statement
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
Supplemental information
  • MIXTURE LIST-Contains : Molybdenum (IV) sulfide

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Molybdenum(IV) sulfide, 98%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.