Learn More
MP Biomedicals™ Alpha-Amylase
From Bacillus subtilis
Brand: MP Biomedicals™ 0210044725
77.20 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Description
- CAS Number: 9000-90-2
- EC Number: 232-565-6
- Activity: 500-1,000 units per mg protein

Specifications
Off-White | |
General procedure for the enzymatic liquefaction | |
Bacillus subtilis | |
alpha-Amylase |
5 to 7 | |
4°C | |
25 g | |
Powder |
Safety and Handling
- alpha-amylase
Signal Word
- Danger
Hazard Category
- Respiratory sensitiser Category 1
Hazard Statement
- H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Precautionary Statement
- P284-¢In case of inadequate ventilation! wear respiratory protection.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : alpha-amylase
Your input is important to us. Please complete this form to provide feedback related to the content on this product.