missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Melford N-Acetyl-D-glucosamine
Product usually ships within 24 hours. Research or further manufacturing use only, not for food or drug use.
Brand: Melford A90510-5.0
This item is not returnable.
View return policy
Description
Product usually ships within 24 hours. Research or further manufacturing use only, not for food or drug use.
CAS Number: 7512-17-6
Molecular Formula: C8H15NO6
Molecular Weight: 221.21
Specifications
7512-17-6 | |
C8H15NO6 | |
5 g |
Product Suggestions
Customers who viewed this item also viewed.
Viewing 1-5 of 7
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Melford N-Acetyl-D-glucosamine > 5 g
Spot an opportunity for improvement?Share a Content Correction