missing translation for 'onlineSavingsMsg'
Learn More
N-Benzyl-3-pyridinemethylamine, 97%, Thermo Scientific Chemicals Product Code.: 11489740

N-Benzyl-3-pyridinemethylamine, 97%, Thermo Scientific Chemicals

Product Code. 11489740
1 g
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy

Product Code. 11489740

Brand: Thermo Scientific Alfa Aesar H50978.03

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 63361-56-8
Molecular Formula C13H14N2
Molecular Weight (g/mol) 198.27
MDL Number MFCD00716866,MFCD08061098
InChI Key COHYOBKZKMKMIX-UHFFFAOYSA-N
Synonym benzyl-pyridin-3-ylmethyl-amine, n-nicotinylbenzylamine, benzyl pyridin-3-ylmethyl amine, benzylpyridin-3-ylmethylamine, 1-phenyl-n-pyridin-3-ylmethyl methylamine, n-benzyl-3-pyridinemethylamine, n-benzyl-1-pyridin-3-yl methanamine, benzyl 3-pyridylmethyl amine, acmc-20aos7, 3-benzylaminomethyl pyridineShow MoreShow Less
PubChem CID 293794
IUPAC Name 1-phenyl-N-(pyridin-3-ylmethyl)methanamine
SMILES C(NCC1=CC=CN=C1)C1=CC=CC=C1

Specifications

Quantity 1 g
Formula Weight 198.27
Percent Purity 97%
Chemical Name or Material N-Benzyl-3-pyridinemethylamine

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

4 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberL02T042 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog NumberH50978.03
Lot NumberJ21Q045 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog NumberH50978.03
Lot NumberT15H053 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog NumberH50978.03
Lot NumberC29W033 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog NumberH50978.03

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
N-Benzyl-3-pyridinemethylamine, 97%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.