missing translation for 'onlineSavingsMsg'
Learn More
n-Butyl isocyanide, 98+%, Thermo Scientific Chemicals Product Code.: 10607132

n-Butyl isocyanide, 98+%, Thermo Scientific Chemicals

Product Code. 10607132
5 g, Glass bottle
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy

Product Code. 10607132

Brand: Thermo Scientific Acros 158380050

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2769-64-4
Molecular Formula C5H9N
Molecular Weight (g/mol) 83.13
MDL Number MFCD00000008
InChI Key FSBLVBBRXSCOKU-UHFFFAOYSA-N
Synonym n-butyl isocyanide, butyl isocyanide, 1-butylisocyanide, n-butylisocyanide, bu-tylisonitrile, n-butylisonitrile, n-butyl isonitrile, butane,1-isocyanoShow MoreShow Less
PubChem CID 76008
IUPAC Name 1-isocyanobutane
SMILES CCCC[N+]#[C-]

Specifications

CAS Min % 98.0
CAS Max % 100.0
Color Colorless to Yellow
Density 0.7900g/mL
Infrared Spectrum Authentic
Assay Percent Range 98+%
Packaging Glass bottle
Linear Formula CH3(CH2)3NC
Refractive Index 1.3950 to 1.397
Quantity 5 g
Fieser 11,99; 14,63
Specific Gravity 0.79
Formula Weight 83.13
Percent Purity 98+%
Physical Form Liquid
Chemical Name or Material n-Butyl isocyanide, 98+%
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0456182 Certificate TypeCertificate of Analysis Date07/09/2023 Catalog Number158380050,158380010
Lot NumberA0454288 Certificate TypeCertificate of Analysis Date25/08/2023 Catalog Number158380050,158380010
Lot NumberA0403910 Certificate TypeCertificate of Analysis Date15/12/2022 Catalog Number158380050,158380010
Lot NumberA0413193 Certificate TypeCertificate of Analysis Date13/12/2022 Catalog Number158380050,158380010
Lot NumberA0443736 Certificate TypeCertificate of Analysis Date15/07/2022 Catalog Number158380050,158380010
compliance-icons compliance-icons
Product Identifier
  • n-Butyl isocyanide
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
  • H373-May cause damage to organs.
  • H331-Toxic if inhaled.
  • H311-Toxic in contact with skin.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
n-Butyl isocyanide, 98+%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.