missing translation for 'onlineSavingsMsg'
Learn More
N'-Hydroxy-2-[(4-methylphenyl)sulfonyl]ethanimidamide, 97%, Thermo Scientific™ Product Code.: 10209542

N'-Hydroxy-2-[(4-methylphenyl)sulfonyl]ethanimidamide, 97%, Thermo Scientific™

Product Code. 10209542
1 g, Amber Glass Bottle
Click to view available options
Quantity:
1 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy

Product Code. 10209542

Brand: Thermo Scientific Maybridge MO01117DA

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Specifications

CAS 72660-98-1
CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Molecular Formula C9H12N2O3S
MDL Number MFCD00169961
Quantity 1 g
Synonym n-hydroxy-2-tosylacetimidamide, n'-hydroxy-2-4-methylphenyl sulfonylethanimidamide, 2-hydroxyimino-1-4-methylphenyl sulfonyl eth-2-ylamine, n'-hydroxy-2-4-methylbenzenesulfonyl ethanimidamide, n'-hydroxy-2-4-methylphenyl sulfonyl ethanimidamide, ethanimidamide,n-hydroxy-2-4-methylphenyl sulfonyl, z-n'-hydroxy-2-4-methylbenzenesulfonyl ethanimidamide, z-n'-hydroxy-2-tosylacetimidamide, hydroxymethylphenylsulfonylethanimidamide
InChI Key ZPEOWHYBZZPAEJ-UHFFFAOYSA-N
SMILES CC1=CC=C(C=C1)S(=O)(=O)CC(N)=NO
IUPAC Name N'-hydroxy-2-(4-methylphenyl)sulfonylethanimidamide
Molecular Weight (g/mol) 228.27
PubChem CID 9582827
Formula Weight 228.27
Percent Purity 97%
Chemical Name or Material N';-Hydroxy-2-[(4-methylphenyl)sulfonyl]ethanimidamide
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
N'-Hydroxy-2-[(4-methylphenyl)sulfonyl]ethanimidamide, 97%, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.