missing translation for 'onlineSavingsMsg'
Learn More
N-Methyl[2-(3-methyl-1,2,4-oxadiazol-5-yl)phenyl]methylamine, 95%, Thermo Scientific™ Product Code.: 10451704

N-Methyl[2-(3-methyl-1,2,4-oxadiazol-5-yl)phenyl]methylamine, 95%, Thermo Scientific™

Product Code. 10451704
250 mg, Amber Glass Bottle
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy

Product Code. 10451704

Brand: Thermo Scientific Maybridge CC49646CB

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 879896-56-7
Molecular Formula C11H13N3O
Molecular Weight (g/mol) 203.25
MDL Number MFCD09702360
InChI Key LJDBGVKOXDSIQQ-UHFFFAOYSA-N
Synonym n-methyl 2-3-methyl-1,2,4-oxadiazol-5-yl phenyl methylamine, benzenemethanamine,n-methyl-2-3-methyl-1,2,4-oxadiazol-5-yl, methyl 2-3-methyl-1,2,4-oxadiazol-5-yl phenyl methyl amine, n-methyl-1-2-3-methyl-1,2,4-oxadiazol-5-yl phenyl methanamine, methyl 2-3-methyl 1,2,4-oxadiazol-5-yl phenyl methyl amineShow MoreShow Less
PubChem CID 24229504
SMILES CNCC1=CC=CC=C1C1=NC(C)=NO1

Specifications

CAS Min % 94.5
CAS Max % 100.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 250 mg
IUPAC Name N-methyl-1-[2-(3-methyl-1,2,4-oxadiazol-5-yl)phenyl]methanamine
Formula Weight 203.24
Percent Purity 95%
Chemical Name or Material N-methyl[2-(3-methyl-1, 2, 4-oxadiazol-5-yl)phenyl]methylamine
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
N-Methyl[2-(3-methyl-1,2,4-oxadiazol-5-yl)phenyl]methylamine, 95%, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.