missing translation for 'onlineSavingsMsg'
Learn More
N,N-Diethylhydroxylamine, 97%, Thermo Scientific Chemicals Product Code.: 10626302

N,N-Diethylhydroxylamine, 97%, Thermo Scientific Chemicals

Product Code. 10626302
100 g, Glass bottle
Click to view available options
Quantity:
5 g
100 g
500 g
Packaging:
Glass bottle
This item is not returnable. View return policy

Product Code. 10626302

Brand: Thermo Scientific Acros 114421000

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3710-84-7
Molecular Formula C4H11NO
Molecular Weight (g/mol) 89.14
MDL Number MFCD00002126
InChI Key FVCOIAYSJZGECG-UHFFFAOYSA-N
Synonym diethylhydroxylamine, ethanamine, n-ethyl-n-hydroxy, n-hydroxydiethylamine, n,n-diethylhydroxyamine, hydroxylamine, n,n-diethyl, unii-314i05edvh, ccris 964, hydroxydiethylamine, diethyl hydroxylamine, diethyl hydroxykamineShow MoreShow Less
PubChem CID 19463
IUPAC Name N,N-diethylhydroxylamine
SMILES CCN(CC)O

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point -26°C to 25°C
Density 0.8670g/mL
Boiling Point 125.0°C to 130.0°C
Flash Point 45°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Linear Formula (C2H5)2NOH
Refractive Index 1.4190 to 1.4220
Quantity 100 g
Beilstein 04, 536
Fieser 06,187
Specific Gravity 0.867
Solubility Information Solubility in water: soluble.
Formula Weight 89.14
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material N, N-Diethylhydroxylamine, 97%
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

1 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0381446 Certificate TypeCertificate of Analysis Date25/08/2023 Catalog Number114425000,114420050,114421000
compliance-icons compliance-icons
Product Identifier
  • N,N-Diethylhydroxylamine
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Flammable liquid Category 3
  • Specific target organ toxicity Category 3
Hazard Statement
  • H411-Toxic to aquatic life with long lasting effects.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
N,N-Diethylhydroxylamine, 97%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.