missing translation for 'onlineSavingsMsg'
Learn More
n-Nonane, 99%, pure, Thermo Scientific Chemicals Product Code.: 10153560

n-Nonane, 99%, pure, Thermo Scientific Chemicals

Product Code. 10153560
100 mL, Glass bottle
Click to view available options
Quantity:
2 mL
100 mL
500 mL
2.5 L
Packaging:
Glass bottle
This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.

Product Code. 10153560

Brand: Thermo Scientific Acros 129111000

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 111-84-2
MDL Number MFCD00009574
InChI Key BKIMMITUMNQMOS-UHFFFAOYSA-N
Synonym n-nonane, shellsol 140, nonyl hydride, nonan, heptane, ethyl, unii-t9w3vh6g10, ccris 6081, hsdb 107, dsstox_cid_5796, nonane, analytical standardShow MoreShow Less
PubChem CID 8141
ChEBI CHEBI:32892
IUPAC Name nonane
SMILES CCCCCCCCC

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point -53.0°C
Color Colorless
Density 0.7180g/mL
Boiling Point 151.0°C
Flash Point 31°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Molecular Formula C9H20
Linear Formula CH3(CH2)7CH3
Refractive Index 1.4040 to 1.4060
Quantity 100 mL
Beilstein 01, 165
Specific Gravity 0.718
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in alcohol, slightly soluble in benzene, acetone, chloroform
Molecular Weight (g/mol) 128.26
Formula Weight 128.26
Percent Purity 99%
Grade Pure
Physical Form Liquid
Chemical Name or Material n-Nonane
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0443432 Certificate TypeCertificate of Analysis Date31/01/2025 Catalog Number129110025,129111000,129115000
Lot NumberA0466212 Certificate TypeCertificate of Analysis Date17/09/2024 Catalog Number129110025,129111000,129115000
Lot NumberA0402059 Certificate TypeCertificate of Analysis Date15/12/2022 Catalog Number129110025,129111000,129110020,129115000
Lot NumberA0418820 Certificate TypeCertificate of Analysis Date28/10/2022 Catalog Number129110025,129111000,129110020,129115000
Lot NumberA0443432 Certificate TypeCertificate of Analysis Date22/06/2022 Catalog Number129110025,129111000,129110020,129115000
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • n-Nonane
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 3
  • Aspiration toxicity Category 1
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
  • LONG-TERM AQUATIC HAZARD Chronic 1
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H304-May be fatal if swallowed and enters airways.
  • H315-Causes skin irritation.
  • H336-May cause drowsiness or dizziness.
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P331-Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
n-Nonane, 99%, pure, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.