Learn More
Thermo Scientific™ Nalgene™ Büchner Two-Piece Polypropylene Funnels
GREENER_CHOICE
Separate the lightweight and durable two-piece polypropylene funnels for easy cleaning. These funnels won't chip, crack or break.
£24.30 - £88.25
Specifications
Material Polypropylene
Autoclavable Autoclavable
View More Specs
Products 5
Product Code Brand Diameter (Metric)Top Capacity (Metric) Price Quantity & Availability  
Product Code Brand Diameter (Metric)Top Capacity (Metric) Price Quantity & Availability  
10657711
GREENER_CHOICE
View Documents Promotion Details
Thermo Scientific™
4280-0550
58 mm 83 mL
£24.30
Each
In Stock
to see stock.
Add to basket
10525492
GREENER_CHOICE
View Documents Promotion Details
Thermo Scientific™
4280-0700
75 mm 186 mL
£29.20
Each
In Stock
to see stock.
Edge
Add to basket
10701411
GREENER_CHOICE
View Documents Promotion Details
Thermo Scientific™
4280-0900
99 mm 358 mL
£49.60
Each
In Stock
to see stock.
Add to basket
10711411
GREENER_CHOICE
View Documents Promotion Details
Thermo Scientific™
4280-1100
132 mm 919 mL
£72.00
Each
In Stock
to see stock.
Edge
Add to basket
10186350
GREENER_CHOICE
View Documents
Thermo Scientific™
4280-1500
175 mm 2.91 L
£88.25
Each
Estimated Shipment: 08-07-2025
to see stock.
Add to basket
Description
Description
Separate the lightweight and durable Thermo Scientific™ Nalgene™ Büchner Two-Piece Polypropylene Funnels for easy cleaning. These autoclavable funnels won't chip, crack or break making them a valuble asset to lab safety programs.
  • Sturdy funnels consist of cup and funnel with perforated plate for easy cleaning and good chemical resistance
  • All sizes are made entirely of polypropylene and are autoclavable to 135°C (275°F)
Recycling Information:
Nalgene reusable labware products are designed to last many years under typical laboratory conditions. Yet, as with all durable products, they need to be retired and replaced eventually.  Thermo Fisher Scientific encourages recycling Nalgene polypropylene (PP) and polypropylene copolymer (PPCO) labware at end of product life in communities where polypropylene can be recycled. Please also recycle the corrugate cartons and any plastic bags (LDPE) included in your product packaging.
Specifications
Specifications
Polypropylene
Autoclavable
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Product Title
Thermo Scientific™ Nalgene™ Büchner Two-Piece Polypropylene Funnels