missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Nalgene™ CleanSheets™ Polyethylene Bench/Drawer Liner
GREENER_CHOICE

Find a variety of uses for these polyethylene bench/drawer liners, such as sink liners or titration backgrounds.

Brand:  Thermo Scientific™ 6283-1850

414.94 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.


 View more versions of this product

Product Code. 11304555

  • £592.00 / Pack of 2
In Stock
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Find a variety of uses for Thermo Scientific™ Nalgene™ CleanSheets™ Polyethylene Bench/Drawer Liners, such as sink liners or titration backgrounds. These liners can be cut to any size and stapled, tacked or taped. They provide cushioning and liquid barrier protection.

  • Made of 0.125″ (3.2mm) thick, closed-cell polyethylene foam to cushion benchtop items
  • Smooth and resilient, this non-absorbent pad is versatile in your lab — use for a multitude of purposes
  • Excellent chemical resistance provides protection against spills wherever placed
  • Can be cut to any size to fit benches, shelves, drawers — anywhere protection is needed against spills, vibration, heat, dirt or breakage
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Bench/Drawer Liner
Non-absorbent
15.2 m
46 cm
2
Liners Clean Sheets XLPE 45.7 cm x 15 m
Polyethylene
50 ft.
18 in.
Polyethylene bench/drawer liners
1
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific™ Nalgene™ CleanSheets™ Polyethylene Bench/Drawer Liner > 18 in. x 50 ft.

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.