missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Nalgene™ LDPE, Round Carboy

For Use With: Store and transport reagents; Closure Material: Polypropylene; Color: White

Brand:  Thermo Scientific™ 2210-0020

70.14 GBP valid until 2025-07-11
Use promo code "25721" to get your promotional price.


Product Code. 10782682

  • £116.00 / Each
In Stock
to see stock.
Edge
Explore more special offers
Add to basket

Description

Description

Store and transport reagents with the Thermo Scientific™ Nalgene™ Round LDPE Carboys, featuring wide shoulder handles that allow for easy carrying and pouring with gloved hands. These carboys accept 13.5 rubber stoppers for an alternative sealing method.

  • Molded-in graduations of 5L and 1 gal. increments
  • Leakproof†

Includes:

White polypropylene screw cap and 83B closure

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Not Autoclavable
10 L
83B
Round LDPE Carboy w/handle
250 mm
389 mm
LDPE
1 Ea.
2.6 gal.
Polypropylene
White
64 mm
Store and transport reagents
Handles
6/Cs.
Carboy with Handle
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number1424929 Certificate TypeCertificate of Compliance Date20/03/2025 Catalog Number2210-0020
Lot Number1420639 Certificate TypeCertificate of Compliance Date08/02/2025 Catalog Number2210-0020
Lot Number1298468 Certificate TypeCertificate of Compliance Date10/12/2024 Catalog Number2210-0020
Lot Number1317666 Certificate TypeCertificate of Compliance Date10/12/2024 Catalog Number2210-0020
Lot Number1417475 Certificate TypeCertificate of Compliance Date26/11/2024 Catalog Number2210-0020
View accessory products
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific™ Nalgene™ LDPE, Round Carboy > 10 L

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

† The term “leakproof” applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for 2 minutes, and no water escapes;. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.