missing translation for 'onlineSavingsMsg'
Learn More
Naltrexone hydrochloride, Thermo Scientific™ Product Code.: 15453339

Naltrexone hydrochloride, Thermo Scientific™

Product Code. 15453339
1g
Click to view available options
Quantity:
250mg
1g
This item is not returnable. View return policy

Product Code. 15453339

Brand: Thermo Scientific Alfa Aesar J60590.03

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

A congener of naloxone and also an opioid antagonist

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
A congener of naloxone and also an opioid antagonist.Naltrexone hydrochloride is used in the treatment of alcohol and utilized for deceasing drinking habit. It is also used to prevent euphorigenic effects in the treatment of patients addicted to opioids. Further, it is used as a specific opiate antagonist.

Solubility
Soluble in water at 50mg/ml.

Notes
Light sensitive. Incompatible with bases and oxidizing agents. Store in a cool place.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 16676-29-2
Molecular Formula C20H24ClNO4
Molecular Weight (g/mol) 377.865
MDL Number MFCD00069324
InChI Key RHBRMCOKKKZVRY-ITLPAZOVSA-N
Synonym naltrexone hydrochloride, naltrexone hcl, trexan, depade, antaxone, revia, nalorex, unii-z6375yw9sf, nemexin
PubChem CID 5485201
IUPAC Name (4R,4aS,7aR,12bS)-3-(cyclopropylmethyl)-4a,9-dihydroxy-2,4,5,6,7a,13-hexahydro-1H-4,12-methanobenzofuro[3,2-e]isoquinoline-7-one;hydrochlorideShow MoreShow Less
SMILES C1CC1CN2CCC34C5C(=O)CCC3(C2CC6=C4C(=C(C=C6)O)O5)O.Cl

Specifications

Quantity 1g
Beilstein 3580333
Sensitivity Light sensitive
Merck Index 14,6363
Solubility Information Soluble in water at 50mg/ml.
Formula Weight 377.86
Chemical Name or Material Naltrexone hydrochloride

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberQ03L004 Certificate TypeCertificate of Analysis Date06/04/2025 Catalog NumberJ60590.MD,J60590.03
Lot NumberS20I007 Certificate TypeCertificate of Analysis Date14/07/2022 Catalog NumberJ60590.MD,J60590.03
Lot NumberE22Z020 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog NumberJ60590.MD,J60590.03
Lot NumberQ08C005 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog NumberJ60590.MD,J60590.03
Lot NumberT07G038 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog NumberJ60590.MD,J60590.03

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Naltrexone hydrochloride, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.