Learn More
Naphthalene, 99+%, Thermo Scientific Chemicals
Description
Naphthalene is formerly important in dyestuff manufacture. It acts as a precursor to phthalic anhydride. Naphthalene is used in the manufacturing of phthalic anhydride, naphthalene sulfonic acids and hydrogenated naphthalenes such as 1,2,3,4-tetrahydronaphthalene, which is used as a precursor for various dyestuffs, pigments, rubber processing chemicals and in pharmaceuticals. Its molten form is used as solvent replacing the high- boiling solvents like dichlorobenzene, nitrobenzene and durene. It is involved as reaction medium for in the Diels-Alder reaction between C60 with anthracene.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Naphthalene is formerly important in dyestuff manufacture. It acts as a precursor to phthalic anhydride. Naphthalene is used in the manufacturing of phthalic anhydride, naphthalene sulfonic acids and hydrogenated naphthalenes such as 1,2,3,4-tetrahydronaphthalene, which is used as a precursor for various dyestuffs, pigments, rubber processing chemicals and in pharmaceuticals. Its molten form is used as solvent replacing the high- boiling solvents like dichlorobenzene, nitrobenzene and durene. It is involved as reaction medium for in the Diels-Alder reaction between C60 with anthracene.
Solubility
Soluble in benzene, toluene, chloroform, carbon disulfide, ether. Insoluble in water.
Notes
Incompatible with strong oxidizing agents.

Specifications
Specifications
Melting Point | 80°C to 82°C |
Density | 0.963 |
Boiling Point | 218°C |
Flash Point | 78°C (172°F) |
Odor | Aromatic |
Refractive Index | 1.5821 |
Quantity | 250 g |
UN Number | UN1334 |
Beilstein | 1421310 |
Merck Index | 14,6370 |
Show More |
Product Suggestions
Customers who viewed this item also viewed
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
A0460958 | Certificate of Analysis | 12/04/2024 | A13188.0B,A13188.30,A13188.0I |
A0460959 | Certificate of Analysis | 12/04/2024 | A13188.0B,A13188.30,A13188.0I |
A0446834 | Certificate of Analysis | 24/10/2023 | A13188.0B,A13188.30,A13188.0I |
A0429345 | Certificate of Analysis | 19/10/2022 | A13188.0B,A13188.30,A13188.0I |
A0429435 | Certificate of Analysis | 19/10/2022 | A13188.0B,A13188.30,A13188.0I |
Safety and Handling




- Naphthalene
- Warning
- Acute toxicity Category 4
- LONG-TERM AQUATIC HAZARD Chronic 1
- ACUTE AQUATIC Acute 1
- Carcinogenicity Category 2
- Flammable solid Category 2
- H228-Flammable solid.
- H302-Harmful if swallowed.
- H351-Suspected of causing cancer.
- H400-Very toxic to aquatic life.
- H410-Very toxic to aquatic life with long lasting effects.
- P201-Obtain special instructions before use.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P264-Wash face, hands and anyexposed skin thoroughly after handling.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
- P370+P378-In case of fire: Use dry sand, dry chemical or alcohol-resistant foam to extinguish.
- MIXTURE LIST-Contains : Naphthalene
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.