Learn More
Thermo Scientific™ National Assembled Standard Opening Vial, Cap and Septum Kits
Thermo Scientific™ National Assembled Standard Opening Vial, Cap and Septum Kits provide convenience, efficiency and stackable storage options.
Brand:  Thermo Scientific™ C4013-19A
 View more versions of this product
Product Code. 11995673
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
Thermo Scientific™ National Assembled Standard Opening Vial, Cap and Septum Kits provide convenience, efficiency and stackable storage options.
  • Packaged in convenient vial trays with clear covers or in economical polybags
  • Vial trays are stackable for easy storage
  • All caps are manufactured from virgin polypropylene unless noted
Includes: 
100 vials with pre-attached caps and septa
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Black
Amber Glass; PP Black Cap; White Virgin PTFE septum; in storage tray
Glass, PTFE
PTFE
Polypropylene
8-425
White
Amber
2 mL
Storage Tray
Polypropylene
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Thermo Scientific™ National Assembled Standard Opening Vial, Cap and Septum Kits > Amber Glass; PP Black Cap; White Virgin PTFE septum; in storage tray