Learn More
Nickel Electro™ Nickel Crucible Lid
Complete a Nickel Crucible with a flat lid that has a depression for centering and an extension on one side that serves as a handle. Nickel Electro™ Nickel Crucible Lid comes in a range of sizes to fit the dimensions of the crucible being used.
Brand: Nickel Electro™ 46
Includes: DiameterMetricOuter
Requires : Autoclavable,dimMaterial,ProductTypeSpecs
Description
- Resistant to corrosion
- Should not be used with nitric acid
- Prevents chance of hot material spattering out of open crucible
- Cost effective replacement for platinum
Crucible lid, nickel, handle
Specifications
Nickel | |
Autoclavable | |
Crucible lid |
Each | |
30 mm |