missing translation for 'onlineSavingsMsg'
Learn More

Nickel Electro™ Nickel Round Bottom Evaporating Basin

Evaporate solutions with a round bottom nickel evaporating dish. Nickel Electro™ Nickel Evaporating Basin with Spout comes in various capacities to accommodate the volume of solution.

Brand:  Nickel Electro™ 718

 View more versions of this product

Product Code. 12757366

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

Explore more special offers

Includes: CapacityMetric,DiameterMetric,HeightMetric

Requires : BottomShape,Material,dimProductType
This item is not returnable. View return policy

Description

Description

  • Crevice free
  • Round bottom
  • Non magnetic
  • Includes pouring spout
  • 99.6% nickel content
  • Supplied in standard packs of 5

Crevice free, round bottom, pouring spout

Specifications

Specifications

Round
Nickel
Evaporating Bowl
100 mm
250 mL
Round
40 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Nickel Electro™ Nickel Round Bottom Evaporating Basin >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.