missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NOB1 Mouse anti-Human, Clone: 8C11A12, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 66048-1-IG-20UL
This item is not returnable.
View return policy
Description
In yeast, over 200 protein and RNA cofactors are required for ribosome assembly, and these are generally conserved in eukaryotes. These factors orchestrate modification and cleavage of the initial 35S precursor rRNA transcript into the mature 18S, 5.8S, and 25S rRNAs, folding of the rRNA, and binding of ribosomal proteins and 5S RNA. Nob1 is involved in pre-rRNA processing. In a late cytoplasmic processing step, Nob1 cleaves a 20S rRNA intermediate at cleavage site D to produce the mature 18S rRNA.Specifications
NOB1 | |
Monoclonal | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q9ULX3 | |
NOB1 | |
NOB1 Fusion Protein Ag17907 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG1 |
Western Blot, Immunohistochemistry (Paraffin) | |
8C11A12 | |
Unconjugated | |
NOB1 | |
ART 4, ART4, MST158, MSTP158, NOB1, NOB1P, Protein ART 4, PSMD8BP1, RNA binding protein NOB1 | |
Mouse | |
Protein G | |
RUO | |
28987 | |
-20°C | |
Liquid |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-4 of
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
NOB1 Mouse anti-Human, Clone: 8C11A12, Proteintech > 20 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction