Learn More
Invitrogen™ Nitrocellulose/Filter Paper Sandwich, 0.2 μm, 8.3 x 7.3 cm
Ideal for blotting proteins and nucleic acids
Brand: Invitrogen™ LC2000
170.64 GBP valid until 2025-05-31
Use promo code "25977" to get your promotional price.
Description
The 0.2μm Pore Size is ideal for the transfer of low molecular weight proteins (less than 20kDa) and nucleic acids (less than 300bp).
- Composed of 100% pure nitrocellulose to provide high-quality transfer
- Contain no support fabrics or detergents
- Are compatible with commonly used transfer conditions and detection methods such as staining, immunodetection, fluorescence, and radiolabeling
- Provide high-sensitivity with low background
- Are supplied in a pre-cut, pre-assembled membrane/filter paper sandwich for convenience
- A protein's properties (i.e., charge, hydrophobicity, etc.) affect its ability to bind to membrane surfaces
DNA and RNA Purification and Analysis, Northern Blotting, Nucleic Acid Gel Electrophoresis and Blotting, Proteins, Expression, Isolation and Analysis, Western Blotting

Specifications
8.3 cm | |
Nitrocellulose Pre-Cut Blotting Membrane, 0.2μm pore size | |
Sandwich | |
0.2 μm | |
Room Temperature | |
0.8 mm |
7.3 cm | |
20 membrane/filter paper sandwiches | |
Nitrocellulose | |
Store at room temperature. | |
7.3 cm x 8.3 cm | |
Novex |
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Special Offers
-
Multiple Manufacturers
Powering Proteomics
Advance your proteomics research with essential toolsPromo Code: 25977Expires: 2025-05-31
New Offer Save Up to 92%
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only. Not for use in diagnostic procedures.