missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Macherey-Nagel™ Nucleogel™ HPLC Column
Brand: Macherey-Nagel™ 719469
This item is not returnable.
View return policy
Specifications
Polymer-based | |
Purification of Peptides, Large Proteins and Oligonucleotides, HPLC | |
8 μm | |
HPLC Column | |
L23 |
Polymer | |
0.1 M Na2SO4 + 0.2 % NaN3 | |
Polymer-based | |
Nucleogel SAX 1000-8 |
Product Suggestions
Customers who viewed this item also viewed.
Viewing 1-4 of
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Macherey-Nagel™ Nucleogel™ HPLC Column > Material: Nucleogel SAX 1000-8; I.D.: 4.6mm
Spot an opportunity for improvement?Share a Content Correction