Learn More
Thermo Scientific™ Nunc™ Cell Factory™ System Accessories
Further the development of applications that require a more closed cell culture system using Thermo Scientific™ Nunc™ Cell Factory™ Accessories.
Brand:  Thermo Scientific™ 140120
182.17 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Product Code. 11316881
  • £213.00 / Pack of 2
Estimated Shipment: 15-09-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
Further the development of applications that require a more closed cell culture system using Thermo Scientific™ Nunc™ Cell Factory™ Accessories. Reduce the need for open interventions and support the use of aseptic methods for fluid handing relating to filling, inoculation, feeding and harvesting of the Cell Factory system.
Connections made easy
  • Plug a media bag or tubing set into any cell factory
Save time and reduce costs
  • Cleaning processes and sterility validations are minimized
  • Most accessories are gamma irradiated and individually wrapped (Note: Connector Cat. Nos- 140099 and 173250 are not available gamma irradiated)
Prevent buildup of back pressure during filling application
  • Venting options provide additional air venting when filling and harvesting by gravity or with a peristaltic pump
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Cell Factory Filling Tubing Set
2
System Accessories
Each
Platinum Cured Silicone Tubing and Polycarbonate Connector
2
Sterile
Gamma Irradiated
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Thermo Scientific™ Nunc™ Cell Factory™ System Accessories > Filling Tubing Set, Irradiated, 2/cs