missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Nunc™ Storage Vials with Caps
GREENER_CHOICE

Assure safe centrifugation and storage of PCR reagents, restriction enzymes, and other diagnostic and biochemical reagents or samples by using these storage vials with caps.

Brand:  Thermo Scientific™ 264262

298.76 GBP valid until 2025-03-28
Use promo code "24107" to get your promotional price.


 View more versions of this product

Product Code. 10694022

  • £388.00 / Pack of 500
In Stock
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Assure safe centrifugation and storage of PCR reagents, restriction enzymes and other biochemical reagents and samples by using Thermo Scientific™ Nunc™ Storage Vials with Caps. Available in 0.5mL and 2.0mL sizes with frosted writing area, the vials have excellent chemical resistance with leakproof closure.

Features:

  • Made from high-purity polypropylene copolymer with excellent chemical resistance
  • Leakproof high-density polyethylene screw closure
  • Centrifuged to a maximum of 13,000 x g
  • Temperature range: -40°C to 95°C One full turn removes the ergonomic closure
  • RNase/DNase-free

Ordering Information:

Racks 264263 and 270972 are compatible with these vials. Color coders available: 254416, 264335, 264408, 264440, 264459, 264467, 264491.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

PPCO
0.9 mL
500
Nunc
Polyethylene
500 Cs.
Storage Vial with Cap
Sterile
500
0.5 mL
0.9 mL
Screw Cap
Certificate of Analysis (COA), Certificate of Conformance (COC)
264263, 270972 Rack
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Schematics & Diagrams

Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number1296815 Certificate TypeCertificate of Compliance Date11/12/2024 Catalog Number264262
Lot Number1337590 Certificate TypeCertificate of Processing Date04/12/2024 Catalog Number264262
Lot Number1400435 Certificate TypeCertificate of Processing Date04/12/2024 Catalog Number264262
Lot Number1286175 Certificate TypeCertificate of Processing Date04/12/2024 Catalog Number264262
Lot Number1382816 Certificate TypeCertificate of Processing Date04/12/2024 Catalog Number264262
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific™ Nunc™ Storage Vials with Caps > Total vol.: 0.9mL; Working vol.: 0.5mL

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.