Learn More
Masterflex™ Nylon Male Luer Fitting
Sterilizable male luer fitting available configured with a variety of fitting options. Cole-Parmer™ Nylon Male Luer Fitting has good chemical resistance.
Brand: Masterflex™ 45505-72
Description
- Good chemical resistance
- Max temperature 455°F (235°C)
- Sterilize by autoclave, ethylene oxide, or gamma irradiation
Specifications
Nylon | |
Fits Male luer x male luer adapter | |
25 Pack | |
Male Luer Fitting |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.