Learn More
Masterflex™ Nylon Sanitary Clamp Fittings
Compatible with Metal and Plastic Tri-Clamp™ Fittings
Brand: Masterflex™ 30508-25
This item is not returnable.
View return policy
Description
- Excellent chemical resistance
- Low leachable characteristics
- Autoclavable
Specifications
Autoclavable | |
Sanitary Clamp | |
Tri-Clamp Fittings | |
White |
Autoclave or Ethylene Oxide (ETO) | |
Nylon | |
2 in. |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-5 of 8
Product Content Correction
Product Title
Masterflex™ Nylon Sanitary Clamp Fittings > 2 in.
Spot an opportunity for improvement?Share a Content Correction