Learn More
Merck O-Ring
Brand: Merck XX4002516
This item is not returnable.
View return policy
Description
Specifications
Viton 2-021 | |
Fluoroelastomer | |
23.52 mm | |
2-021 |
25mm Gas Line Filter Holder | |
5 Pack | |
O-Ring |
Product Suggestions
Customers who viewed this item also viewed.
Viewing 1-4 of
Product Content Correction
Product Title
Merck O-Ring > For use with: Viton 2-021; Quantity: 6/Pk.
Spot an opportunity for improvement?Share a Content Correction