Learn More
Macherey-Nagel™ Octyl Phase Polygosil™ C8
Brand:  Macherey-Nagel™ 711300.100
 View more versions of this product
Product Code. 10276261
  • £1560.00 / 100g
Estimated Shipment: 18-03-2025
to see stock.
Explore more special offers
Add to basket
Specifications
5 μm
Liquid Chromatography
Polygosil C8 Phases
60 Å
100 g
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Macherey-Nagel™ Octyl Phase Polygosil™ C8 > Quantity: 100g; Paritcle Size: 5um