Learn More
Sartorius™ OIML Class E2 Proof Line Leaf Weight without Certificate
Constructed from non-magnetizable aluminum or nickel silver. Sartorius™ OIML Class E2 Leaf Weight without Certificate is higly resistant to corrosion.
£43.55 - £45.55
Product Code | Brand | Mass | Density | Material | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Mass | Density | Material | Price | Quantity & Availability | |||
11343334
|
Sartorius™
YCW0121-AC-00 |
1 mg | 2,650kg/m3 | Aluminum |
£45.55
Each |
Estimated Shipment: 07-01-2025
Log in to see stock. |
|||
11363334
|
Sartorius™
YCW0521-AC-00 |
5 mg | 2,650kg/m3 | Aluminum |
£43.55
Each |
Estimated Shipment: 23-01-2025
Log in to see stock. |
|||
11373334
|
Sartorius™
YCW1121-AC-00 |
10 mg | 8,600kg/m3 | Nickel Silver |
£45.55
Each |
Estimated Shipment: 23-01-2025
Log in to see stock. |
|||
11383334
|
Sartorius™
YCW1221-AC-00 |
20 mg | 8,600kg/m3 | Nickel Silver |
£45.55
Each |
Estimated Shipment: 07-02-2025
Log in to see stock. |
|||
11393334
|
Sartorius™
YCW1521-AC-00 |
50 mg | 8,600kg/m3 | Nickel Silver |
£43.55
Each |
Estimated Shipment: 07-02-2025
Log in to see stock. |
|||
11303344
|
Sartorius™
YCW2121-AC-00 |
100 mg | 8,600kg/m3 | Nickel Silver |
£45.55
Each |
Estimated Shipment: 07-02-2025
Log in to see stock. |
|||
11313344
|
Sartorius™
YCW2221-AC-00 |
200 mg | 8,600kg/m3 | Nickel Silver |
£45.55
Each |
Estimated Shipment: 07-02-2025
Log in to see stock. |
|||
11323344
|
Sartorius™
YCW2521-AC-00 |
500 mg | 8,600kg/m3 | Nickel Silver |
£45.55
Each |
Estimated Shipment: 07-02-2025
Log in to see stock. |
|||
Description
- Milligram weights are provided as leaf weights
- With nominal mass values ranging from 1mg to 5mg are made of aluminum has density of 2,650 kg/m3
- Weights with nominal values of 10mg to 500mg are made of nickel silver has density of 8,600 kg/m3
- Tolerances comply with OIML R111-1
Specifications
Proof Line Weight | |
OIML Class E2 |