Learn More
EVOS™ Olympus Apochromat LWD Objective Lens
For demanding applications, especially capturing color images in white light
Brand:  EVOS™ AMEP4751
Product Code. 15681882
  • £1182.00 / Each
Explore more special offers
Description
Description
  • Has long working distance (LWD) objective that is optimized for imaging of slides, cell culture dishes and flasks, and microtiter plates
  • Compatible with EVOS™ Cell Imaging systems
  • Offers optical performance from visible light to near infrared light
  • Meets needs across spectrum of magnifications and optical specifications
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Olympus Apochromat LWD Objective Lens
6.22mm
0.08
EVOS™ XL Core Imaging System, EVOS FL Color Imaging System, EVOS FL Auto Imaging System, EVOS XL Imaging System, EVOS FL Imaging System
2X
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
EVOS™ Olympus Apochromat LWD Objective Lens > Magnification: 2X; Numerical Aperture: 0.08; Working distance: 6.22mm