Learn More
Diba Omnifit™ BenchMark™ Replacement Frits for Chromatography Column Assemblies
Select from a variety of porosities for your media and chemistry
Brand: Diba 006FR-50-10
Description
Replacement frits are available in PTFE or PE and in a range of pore sizes, enabling you to select the most appropriate bed supports for your application. Match the column ID to select the correct fritSpecifications
Chromatography Column Assemblies | |
10 μm | |
50 mm | |
Replacement Chromatography Frit |
PTFE | |
Universal | |
1.9 in. |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.