Learn More
Macherey-Nagel™ Optima™ 17 GC Capillary Column
£268.00 - £980.00
Specifications
Stationary Phase | Optima™ 17 |
---|---|
USP Type | G1, G2, G38 |
Type | GC Capillary Column |
For Use With (Application) | Steroids Analysis, Pesticide Analysis, Drug Analysis |
Temperature Range | 320°C to 340°C |
Product Code | Brand | Length (Metric) | Diameter (Metric) Inner | Film Thickness | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Length (Metric) | Diameter (Metric) Inner | Film Thickness | Price | Quantity & Availability | ||||
10610352
|
Macherey-Nagel™
726848.10 |
10 m | 0.1 mm | 0.1 μm |
£287.00
Each |
Estimated Shipment: 06-06-2025
Log in to see stock. |
||||
10736322
|
Macherey-Nagel™
726747.10 |
10 m | 0.53 mm | 1 μm |
£295.00
Each |
Estimated Shipment: 06-06-2025
Log in to see stock. |
||||
10400902
|
Macherey-Nagel™
726747.15 |
15 m | 0.53 mm | 1 μm |
£356.00
Each |
Estimated Shipment: 06-06-2025
Log in to see stock. |
||||
10596851
|
Macherey-Nagel™
726065.12 |
12 m | 0.2 mm | 0.2 μm |
£268.00
Each |
Estimated Shipment: 06-06-2025
Log in to see stock. |
||||
10747852
|
Macherey-Nagel™
726742.25 |
25 m | 0.25 mm | 0.15 μm |
£515.00
Each |
Estimated Shipment: 06-06-2025
Log in to see stock. |
||||
10194651
|
Macherey-Nagel™
726022.15 |
15 m | 0.25 mm | 0.25 μm |
£319.00
Each |
Estimated Shipment: 06-06-2025
Log in to see stock. |
||||
10174891
|
Macherey-Nagel™
726065.25 |
25 m | 0.2 mm | 0.2 μm |
£515.00
Each |
Estimated Shipment: 06-06-2025
Log in to see stock. |
||||
10012981
|
Macherey-Nagel™
726067.25 |
25 m | 0.25 mm | 0.5 μm |
£515.00
Each |
Estimated Shipment: 06-06-2025
Log in to see stock. |
||||
10608621
|
Macherey-Nagel™
726022.25 |
25 m | 0.25 mm | 0.25 μm |
£515.00
Each |
Estimated Shipment: 06-06-2025
Log in to see stock. |
||||
10082961
|
Macherey-Nagel™
726066.25 |
25 m | 0.2 mm | 0.5 μm |
£515.00
Each |
Estimated Shipment: 06-06-2025
Log in to see stock. |
||||
10639191
|
Macherey-Nagel™
726351.25 |
25 m | 0.32 mm | 0.25 μm |
£551.00
Each |
Estimated Shipment: 06-06-2025
Log in to see stock. |
||||
10409171
|
Macherey-Nagel™
726067.30 |
30 m | 0.25 mm | 0.5 μm |
£515.00
Each |
Estimated Shipment: 06-06-2025
Log in to see stock. |
||||
10276221
|
Macherey-Nagel™
726022.30 |
30 m | 0.25 mm | 0.25 μm |
£515.00
Each |
In Stock
Log in to see stock. |
||||
10479551
|
Macherey-Nagel™
726757.25 |
25 m | 0.32 mm | 0.35 μm |
£551.00
Each |
Estimated Shipment: 06-06-2025
Log in to see stock. |
||||
10689571
|
Macherey-Nagel™
726351.30 |
30 m | 0.32 mm | 0.25 μm |
£551.00
Each |
In Stock
Log in to see stock. |
||||
10226471
|
Macherey-Nagel™
726747.25 |
25 m | 0.53 mm | 1 μm |
£637.00
Each |
Estimated Shipment: 06-06-2025
Log in to see stock. |
||||
10042951
|
Macherey-Nagel™
726744.25 |
25 m | 0.32 mm | 0.5 μm |
£551.00
Each |
Estimated Shipment: 06-06-2025
Log in to see stock. |
||||
10266221
|
Macherey-Nagel™
726742.30 |
30 m | 0.25 mm | 0.15 μm |
£515.00
Each |
Estimated Shipment: 06-06-2025
Log in to see stock. |
||||
10358451
|
Macherey-Nagel™
726755.30 |
30 m | 0.32 mm | 0.15 μm |
£551.00
Each |
Estimated Shipment: 06-06-2025
Log in to see stock. |
||||
10786702
|
Macherey-Nagel™
726757.30 |
30 m | 0.32 mm | 0.35 μm |
£551.00
Each |
Estimated Shipment: 06-06-2025
Log in to see stock. |
||||
10236471
|
Macherey-Nagel™
726747.30 |
30 m | 0.53 mm | 1 μm |
£637.00
Each |
Estimated Shipment: 06-06-2025
Log in to see stock. |
||||
10431852
|
Macherey-Nagel™
726744.30 |
30 m | 0.32 mm | 0.5 μm |
£551.00
Each |
Estimated Shipment: 06-06-2025
Log in to see stock. |
||||
10748042
|
Macherey-Nagel™
726742.50 |
50 m | 0.25 mm | 0.15 μm |
£854.00
Each |
Estimated Shipment: 06-06-2025
Log in to see stock. |
||||
10421662
|
Macherey-Nagel™
726022.50 |
50 m | 0.25 mm | 0.25 μm |
£854.00
Each |
Estimated Shipment: 06-06-2025
Log in to see stock. |
||||
10256561
|
Macherey-Nagel™
726067.50 |
50 m | 0.25 mm | 0.5 μm |
£854.00
Each |
Estimated Shipment: 06-06-2025
Log in to see stock. |
||||
10556511
|
Macherey-Nagel™
726065.50 |
50 m | 0.2 mm | 0.2 μm |
£854.00
Each |
Estimated Shipment: 06-06-2025
Log in to see stock. |
||||
10338451
|
Macherey-Nagel™
726066.50 |
50 m | 0.2 mm | 0.5 μm |
£854.00
Each |
Estimated Shipment: 06-06-2025
Log in to see stock. |
||||
10787082
|
Macherey-Nagel™
726351.50 |
50 m | 0.32 mm | 0.25 μm |
£924.00
Each |
Estimated Shipment: 06-06-2025
Log in to see stock. |
||||
10296561
|
Macherey-Nagel™
726757.50 |
50 m | 0.32 mm | 0.35 μm |
£924.00
Each |
Estimated Shipment: 06-06-2025
Log in to see stock. |
||||
10184941
|
Macherey-Nagel™
726744.50 |
50 m | 0.32 mm | 0.5 μm |
£924.00
Each |
Estimated Shipment: 06-06-2025
Log in to see stock. |
||||
10536501
|
Macherey-Nagel™
726022.60 |
60 m | 0.25 mm | 0.25 μm |
£889.00
Each |
Estimated Shipment: 06-06-2025
Log in to see stock. |
||||
10576511
|
Macherey-Nagel™
726067.60 |
60 m | 0.25 mm | 0.5 μm |
£889.00
Each |
Estimated Shipment: 06-06-2025
Log in to see stock. |
||||
10481272
|
Macherey-Nagel™
726742.60 |
60 m | 0.25 mm | 0.15 μm |
£889.00
Each |
Estimated Shipment: 06-06-2025
Log in to see stock. |
||||
10318261
|
Macherey-Nagel™
726351.60 |
60 m | 0.32 mm | 0.25 μm |
£980.00
Each |
Estimated Shipment: 06-06-2025
Log in to see stock. |
||||
10595371
|
Macherey-Nagel™
726757.60 |
60 m | 0.32 mm | 0.35 μm |
£980.00
Each |
Estimated Shipment: 06-06-2025
Log in to see stock. |
||||
10586691
|
Macherey-Nagel™
726744.60 |
60 m | 0.32 mm | 0.5 μm |
£980.00
Each |
Estimated Shipment: 06-06-2025
Log in to see stock. |
||||
Specifications
Optima™ 17 | |
GC Capillary Column | |
320°C to 340°C |
G1, G2, G38 | |
Steroids Analysis, Pesticide Analysis, Drug Analysis | |
1 Each |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Product Title
Macherey-Nagel™ Optima™ 17 GC Capillary Column