Learn More
Thermo Scientific™ Orion™ AQUAfast™ IV Chemistry Reagents
Minimize the risks of improper measurement and contamination with these chemistry reagents, which are contained in self-filling vials.
Brand: Thermo Scientific™ AC4078
Description
Minimize the risks of improper measurement and contamination with Thermo Scientific™ Orion™ AQUAfast™ IV Chemistry Reagents, which are contained in self-filling vials. Use these reagents with the Orion™ AQUAfast AQ4000 colorimeter to simplify sample preparation and tests. The auto-test ampoules contain a pre-measured amount of liquid reagent and easily fill with the correct volume of sample for simple and fast sample and reagent mixing. Each ampoule contains an Auto-ID barcode that is read by the AQ4000 colorimeter and used to automatically identify the test parameter and initiates the test.
- No. of test: 30
- Consistency and reproducibility improved with AQUAfast ampoules and an AQ4000 meter
- Thermo Scientific™ Auto-ID™ eliminates measurement errors
Specifications
Iron | |
Orion AQUAfast AQ4000 Colorimeter | |
30 Auto-ID Ampoules | |
Colorimetric water and wastewater testing equipment | |
0.2 to 6 mg/L Fe | |
30 Ampoules |
Ampoule | |
AQUAfast iron (total & soluble) Auto-ID ampoules, 30 pack | |
Phenanthroline | |
AQUAfast Zero Vial Auto-Test Kit (Cat. No. AQ4ZER) | |
Wastewater: Standard Methods 3500-Fe-1997 |
Product Suggestions
Customers who viewed this item also viewed
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.