missing translation for 'onlineSavingsMsg'
Learn More
Oxcarbazepine, Tocris Bioscience™ Product Code.: 18075712

Oxcarbazepine, Tocris Bioscience™

Product Code. 18075712
50 mg
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy

Product Code. 18075712

Brand: Tocris Bioscience 3864/50

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Anticonvulsant; inhibits Na+ channel activity

Anticonvulsant; protects mice and rats against generalized tonic-clonic seizures induced by electroshock. Thought to act via inhibition of sodium channel activity.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 28721-07-5
Molecular Formula C15H12N2O2
Molecular Weight (g/mol) 252.273
InChI Key CTRLABGOLIVAIY-UHFFFAOYSA-N
Synonym oxcarbazepine, trileptal, oxcarbamazepine, oxacarbazepine, oxcarbazepina, oxcarbazepinum, timox, oxcarbazepinum inn-latin, oxcarbazepina inn-spanish, oxcarbazepine innShow MoreShow Less
PubChem CID 34312
ChEBI CHEBI:7824
IUPAC Name 5-oxo-6H-benzo[b][1]benzazepine-11-carboxamide
SMILES C1C2=CC=CC=C2N(C3=CC=CC=C3C1=O)C(=O)N

Specifications

Quantity 50 mg
Formula Weight 252.27
Percent Purity >98%
Chemical Name or Material Oxcarbazepine
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Oxcarbazepine, Tocris Bioscience™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.