missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Oxoid™ Baird-Parker Agar

Easily isolate and enumerate Staphylococcus aureus with highly selective and sensitive, ready-to-use media.

Brand:  Thermo Scientific™ PO0168A

18.46 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.


 View more versions of this product

Product Code. 12972588

  • £38.35 / Pack of 10

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description
Show More
Specifications

Specifications

2°C to 10°C
United Kingdom
Agar
Wrapped in single cellulose-based film wrap
90mm Monoplate
Baird-Parker Agar
19.5g ± 1g
10/Pk.
Product Suggestions

Product Suggestions

Videos
SDS
Documents Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number4497243 Certificate TypeCertificate of Analysis Date10/03/2025 Catalog NumberPO0168A
Lot Number4492479 Certificate TypeCertificate of Analysis Date24/02/2025 Catalog NumberPO0168A
Lot Number4491786 Certificate TypeCertificate of Analysis Date18/02/2025 Catalog NumberPO0168A
Lot Number4489467 Certificate TypeCertificate of Analysis Date11/02/2025 Catalog NumberPO0168A
Lot Number4487161 Certificate TypeCertificate of Analysis Date27/01/2025 Catalog NumberPO0168A
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific™ Oxoid™ Baird-Parker Agar >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.