missing translation for 'onlineSavingsMsg'
Learn More

Kartell™ Plastilab™ Polypropylene Pipette Jar

Versatile rugged jar for storing pipettes.

Brand:  Kartell™ 0059200

132.87 GBP valid until 2025-04-13
Use promo code "24122" to get your promotional price.


Product Code. 10642132

  • £219.00 / Each
Estimated Shipment: 02-05-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Versatile rugged jar for storing pipettes.

  • Polypropylene provides cushioning effect for pipettes
  • Tight fitting cover protects during transportation
  • Welded flanged base for stability
  • Can be cut to suit shorter-length pipettes
  • Can be canted on side for drainage
  • Jar autoclavable at 120°C
Specifications

Specifications

Autoclavable
Polypropylene
Pipette Jar
Storing pipettes
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Kartell™ Plastilab™ Polypropylene Pipette Jar > Pipet Jar

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.