missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Oxoid™ Corn Meal Mycological Agar

Maintain the fungal stock culture and observe chlamydispore production by Candida albicans with this mycological medium

Brand:  Thermo Scientific™ CM0103B

267.47 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.


Product Code. 10505753

  • £493.00 / 500g

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Maintain the fungal stock culture and observe chlamydospore production by Candida albicans with Thermo Scientific™ Oxoid™ Corn Meal Mycological Agar (Dehydrated). Microscopic examination of Candida albicans on this medium shows the characteristic chlamydospore production which is an accepted criterion for the identification of this species. Corn Meal Mycological Agar, being a nutritionally impoverished medium, can be employed for the maintenance of stock cultures of fungi, especially the black pigmented varieties.

Use Corn Meal Mycological Agar for chlamydospore production by Candida albicans for the maintenance of fungal stock cultures.

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Corn Meal Agar
Agar
For 29.4L medium
Powder
500 g
Product Suggestions

Product Suggestions

Videos
SDS
Documents Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number6210058 Certificate TypeCertificate of Analysis Date21/01/2025 Catalog NumberCM0103B
Lot Number6206062 Certificate TypeCertificate of Analysis Date04/01/2025 Catalog NumberCM0103B
Lot Number6185739 Certificate TypeCertificate of Analysis Date22/11/2024 Catalog NumberCM0103B
Lot Number6146478 Certificate TypeCertificate of Analysis Date07/09/2024 Catalog NumberCM0103B
Lot Number6114438 Certificate TypeCertificate of Analysis Date06/07/2024 Catalog NumberCM0103B
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific™ Oxoid™ Corn Meal Mycological Agar > 500g

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Laboratory Use Only