missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ M17 Broth (Dehydrated)

Cultivate lactic streptococci and their bacteriophages with this medium

Brand:  Thermo Scientific™ CM0817B

Product Code. 10739393

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

Explore more special offers
This item is not returnable. View return policy

Description

Description

Cultivate lactic streptococci and their bacteriophages with Thermo Scientific™ Oxoid™ M17 Broth (Dehydrated). The use of M17 Broth in conjunction with M17 Agar is recommended by Terzaghi and Sandine1 for bacteriophage assays.

Lactic streptococci are nutritionally fastidious and require complex media for optimal growth2,3. Their homofermentative acid-producing nature require that the medium is well buffered so that the culture pH is maintained above 5.7 during active growth. This maintenance of the pH is important as lower pH can results in injury and reduced recovery of lactic streptococci.

M17 Broth contains di-sodium glycerophoshate which has sufficient buffering capacity to maintain the pH above 5.7 of actively growing cultures even after 24 hours at 30°C. This buffering agent also allows the addition of calcium without a precipitation complex being formed. The calcium-containing medium is used for the assay of bacteriophages of lactic streptococci1.

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Industrial Reference: IDF.
Powder
500 g
M17 Broth
Dehydrated Culture Media
For 13.4L medium
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number3522536 Certificate TypeCertificate of Analysis Date03/09/2022 Catalog NumberCM0817B
Lot Number3446836 Certificate TypeCertificate of Analysis Date13/04/2022 Catalog NumberCM0817B
Lot Number3419164 Certificate TypeCertificate of Analysis Date09/02/2022 Catalog NumberCM0817B
Lot Number3336500 Certificate TypeCertificate of Analysis Date06/10/2021 Catalog NumberCM0817B
Lot Number3335473 Certificate TypeCertificate of Analysis Date06/10/2021 Catalog NumberCM0817B
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific™ M17 Broth (Dehydrated) > M17 Broth; 500g

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Laboratory Use Only