missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Maximum Recovery Diluent (Dehydrated)

Achieve maximum recovery of microorganisms during dilution with this protective, isotonic diluent.

Brand:  Thermo Scientific™ CM0733B

41.45 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.


Product Code. 10229682

  • £87.30 / 500g
In Stock
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Achieve maximum recovery of microorganisms during dilution with this protective, isotonic Thermo Scientific™ Oxoid™ Maximum Recovery Diluent (Dehydrated).

Maximum Recovery Diluent combines the protective effect of peptone in the diluting solution1 with the osmotic support of physiological saline1,2. The low peptone concentration in the diluent reduces organism multiplication in the sample for at least 45 minutes during the dilution stage1,2

Maintain the viability of organisms during dilution process with Maximum Recovery Diluent with low levels of peptone.

  • Allows accurate quantitative procedures to be performed with minimal reduction in viable count.
  • Reduces organism multiplication in the sample for at least one hour during the dilution stage.

The presence of peptone in the diluent provides viable counts to ensure accurate quantitative procedures are performed. The isotonic strength of the diluent ensures recovery of organisms from various sources which may be vulnerable in distilled water or aqueous suspensions.

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Industrial Reference: AFNOR, BSI, EP, IDF, ISO, NMKL
Powder
500 g
Mannitol Salt Agar
Dehydrated Culture Media
For 52.6L medium
Product Suggestions

Product Suggestions

Videos
SDS
Documents Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number6224132 Certificate TypeCertificate of Analysis Date26/02/2025 Catalog NumberCM0733B
Lot Number6216195 Certificate TypeCertificate of Analysis Date07/02/2025 Catalog NumberCM0733B
Lot Number6196872 Certificate TypeCertificate of Analysis Date03/12/2024 Catalog NumberCM0733B
Lot Number6174049 Certificate TypeCertificate of Analysis Date17/10/2024 Catalog NumberCM0733B
Lot Number6151981 Certificate TypeCertificate of Analysis Date21/08/2024 Catalog NumberCM0733B
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific™ Maximum Recovery Diluent (Dehydrated) > Max. Recovery Diluent (MRD, Peptone Salt Broth); 500g

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For In Vitro Diagnostic Use