missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sartorius Vivaspin™ Turbo 4 Ultrafiltration Unit
Handles up to 4mL sample volume in 4 swing-out and fixed angle rotors accepting 15mL centrifuge tubes. Sartorius Vivaspin Turbo 4 Ultrafiltration Unit, features twin vertical membranes and sleek internal design for fastest sample concentration with highest recoveries.
£312.00 - £931.00
Specifications
Description | Turbo 4 Ultrafiltration Unit |
---|---|
Material (Membrane) | PES |
For Use With (Application) | Concentration, Desalting, buffer exchange |
Max. Force | Swing-out rotor : 4000 x gAngle rotor : 7500 x g |
Sample Type | Sample type: Nanoparticles; Peptide; Protein |
Product Code | Brand | MWCO | Quantity | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | MWCO | Quantity | Price | Quantity & Availability | ||||
15448811
|
Sartorius
VS04T91 |
3 kDa | 25/Pk. |
£335.00
Pack of 25 |
In Stock
Log in to see stock. |
||||
15458811
|
Sartorius
VS04T92 |
3 kDa | 100/Pk. |
£920.00
Pack of 100 |
In Stock
Log in to see stock. |
||||
15321335
|
Sartorius
VS04T11 |
5 kDa | 25/Pk. |
£319.00
Pack of 25 |
In Stock
Log in to see stock. |
||||
15354882
|
Sartorius
VS04T12 |
5 kDa | 100/Pk. |
£931.00
Pack of 100 |
In Stock
Log in to see stock. |
||||
16616752
|
Sartorius
VS04T01 |
10 kDa | 25/Pk. |
£590.00
Pack of 25 |
In Stock
Log in to see stock. |
||||
16231961
|
Sartorius
VS04T01 |
10 kDa | 25/Pk. | ||||||
15498801
|
Sartorius
VS04T02 |
10 kDa | 100/Pk. |
£920.00
Pack of 100 |
In Stock
Log in to see stock. |
||||
16636752
|
Sartorius
VS04T02 |
10 kDa | 100/Pk. | ||||||
15367016
|
Sartorius
VS04T21 |
30 kDa | 25/Pk. |
£312.00
Each |
In Stock
Log in to see stock. |
||||
15378826
|
Sartorius
VS04T22 |
30 kDa | 100/Pk. |
£920.00
Pack of 100 |
In Stock
Log in to see stock. |
||||
15311091
|
Sartorius
VS04T31 |
50 kDa | 25/Pk. |
£318.00
Pack of 25 |
In Stock
Log in to see stock. |
||||
15395343
|
Sartorius
VS04T32 |
50 kDa | 100/Pk. |
£920.00
Pack of 100 |
In Stock
Log in to see stock. |
||||
15391577
|
Sartorius
VS04T41 |
100 kDa | 25/Pk. |
£318.00
Pack of 25 |
In Stock
Log in to see stock. |
||||
15418811
|
Sartorius
VS04T42 |
100 kDa | 100/Pk. |
£901.00
Pack of 100 |
In Stock
Log in to see stock. |
||||
Description
Handles up to 4mL sample volume in 4 swing-out and fixed angle rotors accepting 15mL centrifuge tubes. Sartorius Vivaspin Turbo 4 Ultrafiltration Unit, features twin vertical membranes and sleek internal design for fastest sample concentration with highest recoveries.
- High flux PES membrane
- Retentate recovery >95% x pH 1-14 suited polystyrene housing with high chemical compatibility
- Universal rotor compatibility
- Volume graduation for easy detection / monitoring of the concentrate volume
- UV joining technology allows for smooth joint transition between membrane and plastic housing, allowing collection of complete concentrated sample into unique pipette friendly dead stop pocket
Specifications
Turbo 4 Ultrafiltration Unit | |
Concentration, Desalting, buffer exchange | |
Sample type: Nanoparticles; Peptide; Protein | |
2 to 4 mL |
PES | |
Swing-out rotor : 4000 x gAngle rotor : 7500 x g | |
Centrifugal Unit | |
1225 mm |