Learn More
Palladium standard solution, for AAS, 1 mg/ml Pd in 10% HCl, Thermo Scientific Chemicals
Brand: Thermo Scientific Acros 196171000
38.38 GBP valid until 2025-05-31
Use promo code "24127" to get your promotional price.
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For atomic adsorption spectroscopy
Specifications
Palladium standard solution | |
7440-05-3, 7647-01-0, 7732-18-5 | |
100 mL | |
1.05 | |
MFCD00011167 MFCD03457879 MFCD03427452 MFCD03613602 MFCD03613603 MFCD03613604 MFCD03613605 MFCD06798745 | |
997 to 1003 μg/mL of Pd (2+) in 10% HCl (20°C) | |
KDLHZDBZIXYQEI-UHFFFAOYSA-N | |
107.0°C | |
palladium | |
106.42 | |
CHEBI:33363 |
1 mg/ml Pd in 10% HCl | |
Plastic bottle | |
1.0500g/mL | |
Pd | |
Solution | |
Brown/Red | |
-25.0°C | |
[Pd] | |
Soluble in water | |
23938 | |
AAS |
Product Suggestions
Customers who viewed this item also viewed.
Safety and Handling
- Palladium atomic absorption standard solution
Signal Word
- Warning
Hazard Category
- Serious eye damage/eye irritation Category 2
- Substances and mixtures corrosive to metals Category 1
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H290-May be corrosive to metals.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P332+P313-If skin irritation occurs: Get medical advice/attention.
- P337+P313-If eye irritation persists: Get medical advice/attention.
- P390-Absorb spillage to prevent material damage.
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
A0461031 | Certificate of Analysis | 14/03/2024 | 196175000,196171000 |
A0451240 | Certificate of Analysis | 11/03/2024 | 196175000,196171000 |
A0426720 | Certificate of Analysis | 02/02/2023 | 196175000,196171000 |
A0393683 | Certificate of Analysis | 12/12/2022 | 196175000,196171000 |
A0403563 | Certificate of Analysis | 13/12/2021 | 196175000,196171000 |
Special Offers
-
Multiple Manufacturers
Chromatography and Chemicals Promotion
Explore our new promotion and get up to 70% off some of our most popular products.Promo Code: 24127Expires: 2025-05-31
Days
42Hours
12Minutes
37Seconds
51New Offer Save Up to 70%
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
RUO – Research Use Only