missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Honeywell™ Panoramasque™ Full-Face Air Purifying Respirator (APR)
The Panoramasque™ Full-Face APR is a Mono filter with Rd40 thread connection system that offers respiratory protection, depending on the canister type assembled on it.
Brand: Honeywell™ 1710395
View more versions of this product
Description
- Product Type: Full-Face Air Purifying Respirator (APR)
- Material (Body): EPDM or Silicone
- Material (Lens): PMMA, Glass, Chlorine Hazard Specific
- Color: Black, Yellow
- Cartridge: Single Filter via Rd40 (1/7 in.) Connection
- Use with: EN141 or EN143 Class 1 or 2 Cert. Honeywell Filters
- PPE Class: Variable (Dependant on Cartridge)
- Hazard Types: Particulates, Gas, Vapor, Smoke
- Applications: Chemical Industry, Oil and Gas Industries, Pharmaceutical Industry

Specifications
EN 136:1989 | |
Black | |
Full-Face Air Purifying Respirator (APR) | |
Chemical Industry, Oil and Gas Industries, Pharmaceutical Industry | |
Five-Point Harness | |
PMMA |
1/7 in. Rd40 Thread System | |
Mono-Filter | |
EPDM | |
EN141 or EN143 Class 1 or 2 Cert. Honeywell Filters | |
With Speech Diaphragm (Metal) | |
Yes |
Product Suggestions
Customers who viewed this item also viewed.
Viewing 1-4 of
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Honeywell™ Panoramasque™ Full-Face Air Purifying Respirator (APR) > Facepiece Material: EPDM; Material (Lens): PMMA; Color: Black; Includes: Speech Diaphragm (Metal)
Spot an opportunity for improvement?Share a Content Correction