Learn More
Novus Biologicals™ Peanut Agglutinin [Cy5]
Brand:  Novus Biologicals™ CL-1075-NB
Product Code. 18371468
  • £306.00 / 1mg
Explore more special offers
Specifications
Peanut Agglutinin
1 mg
A precipitate may form during storage. This does not have a significant adverse effect on the product. If a precipitate forms upon long-term storage at 4°C, warm to 37C and centrifuge before use. The recommended concentration range for use is 5-20 μg/mL.
1 mg/mL
Store at 4°C in the dark.
10 mM HEPES, 0.15 M NaCl, pH 7.5, 0.1 mM CaCl2, proprietary stabilizer
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Novus Biologicals™ Peanut Agglutinin [Cy5] > 1 mg