Learn More
Sampling Systems™ Pendulum Beaker
These pendulum beakers automatically swing to the horizontal position, meaning that your water samples are less likely to spill when using the TeleScoop.
Brand:  Sampling Systems™ 5065C-502
 View more versions of this product
Product Code. 17902122
  • £33.88 / Each
Estimated Shipment: 06-05-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Telescopic samplers are often used for taking surface samples from rivers, tanks and lakes
  • A wide choice of attachments is available to make the TeleScoop ideally suited for most applications
  • Attachments are interchangeable and simply click into position
Specifications
Specifications
Swabbing Template
Autoclavable
Reusable
Non-sterile
Polypropylene
500 mL
No
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Sampling Systems™ Pendulum Beaker > Pendulum Beaker; Capacity (Metric): 500 mL; Graduated: No; Material: Polypropylene