missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ Lysing Matrix A, 2 mL Tubes

For all sample types except soil for the subsequent isolation of genomic DNA. Lysing Matrix A impact-resistant tubes contain garnet matrix and one 1/4 in. ceramic sphere.

Brand:  MP Biomedicals™ 116910050

279.66 GBP valid until 2025-12-31
Use promo code "24339" to get your promotional price.


 View more versions of this product

Product Code. 12719670

  • £328.00 / Pack of 50

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Tube Size: 2mL
  • Sample Type: Animal tissues, plants, bacteria, yeast, fungi, algae, insects, nematodes
  • Molecule to be Enriched: DNA
  • Grinding matrix is chemically inert and will not bind nucleic acids
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Garnet Matrix, 1/4 in. Ceramic Sphere
Animal Tissues, Plants, Insects, Worms, Bacteria, Yeast, Fungi, Algae
FastPrep instruments or other bead beaters
Lysing Matrix A
Rnase-free
Store at Room Temperature.
Dnase-free
Nucleic Acid Isolation and Purification, Homogenization
50 x 2 mL Tubes
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
MP Biomedicals™ Lysing Matrix A, 2 mL Tubes > Lysing Matrix A; Quantity: 50 x 2 mL Tubes

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.