missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ PureLink™ Genomic DNA Mini Kit

The PureLink™ Genomic DNA Mini Kit enables high-yield, high-purity, genomic DNA (gDNA) extractions from a wide variety of sample types. Use this single kit for genomic DNA purification from blood, tissues, cells, bacteria, swabs, and blood spots, with a familiar silica-based, microcentrifuge spin-column format.

Brand:  Invitrogen™ K182001


 View more versions of this product

Product Code. 10053293

  • £150.00 / Each

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers

For Research Use Only. All usage must comply with product instructions.
 
This item is not returnable. View return policy

Description

Description

The PureLink™ Genomic DNA Mini Kit enables high-yield, high-purity, genomic DNA (gDNA) extractions from a wide variety of sample types. Use this single kit for genomic DNA purification from blood, tissues, cells, bacteria, swabs, and blood spots, with a familiar silica-based, microcentrifuge spin-column format. Features of the PureLink™ Genomic DNA Mini Kit include:

Kit flexibility—one kit works with a variety of sample types and sizes
Ultraclean gDNA—minimal contamination of DNA product means successful downstream application
Improved design—optimized spin-column design and buffer formulation for better yield and purity

Multiple sample sources, one kit
The PureLink™ Genomic DNA Mini Kit can be used with many different sample types, each with its own optimized protocol outlined in the manual.

Optimized components and protocols
The PureLink™ Genomic DNA Mini Kit includes parts and protocols that improve upon the easy and familiar spin-column methods currently in use. Columns are newly configured to bind and release DNA more efficiently. Each sample source uses a specialized lysis procedure and an optimized lysis buffer formulated to enhance proteinase K activity and eliminate protein contamination. A chaotropic salt buffer promotes stable binding of DNA to the column resin, and powerful wash buffers remove all traces of protein and salt. DNA is eluted in a low-salt buffer to allow for pH stabilization of the DNA in storage.

Higher yields and purity
With PureLink™ Genomic DNA Kits you can expect high yields of high-purity gDNA (determined by A260/A280 measurements), no matter the sample type, from bacteria to tissues to blood to cultured cells. The enhanced PureLink™ Lysis and Wash Buffers help to consistently deliver purified gDNA samples with absorbance reading ratios of ∼1.9 for both A260/A280 and A260/A230, indicating no detectable contamination from carryover of protein or guanidine.

Order Info

Shipping Condition: Room Temperature

TRUSTED_SUSTAINABILITY
Specifications

Specifications

• 10 mL PureLink™ Genomic Lysis/Binding Buffer
• 9 mL PureLink™ Genomic Digestion Buffer
• 10 mL PureLink™ Genomic Wash Buffer 1
• 7.5 mL PureLink™ Genomic Wash Buffer 2
• 10 mL PureLink™ Genomic Elution Buffer
• 1 mL RNase A (20 mg/mL)
• 1 mL Proteinase K (20 mg/mL)
• 50 each PureLink™ Spin Columns with Collection Tubes
• 100 PureLink™ Collecion Tubes (2.0 mL)

This kit contains enough reagents for 50 DNA preps. Store all components at room temperature. For long-term storage, Proteinase K and RNase A may be stored at 4°C.
Mini
Genotyping, Real-Time Quantitative PCR (qPCR), Southern Blotting, Sequencing, PCR
100 μL to 1 mL Blood Samples (Medium-Scale)
Genomic DNA Mini Kit
Room Temperature
Silica Spin Column
Bacteria, Blood, Cells, Tissue
Not High-throughput Compatible (Manual)
PureLink™
50 Preps
Genomic DNA
Product Suggestions

Product Suggestions

Videos
Safety and Handling
Show More
SDS
Documents

Documents

Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number3033831 Certificate TypeCertificate of Analysis Date31/01/2025 Catalog NumberK182001
Lot Number3059074 Certificate TypeCertificate of Analysis Date31/01/2025 Catalog NumberK182001
Lot Number2996832 Certificate TypeCertificate of Analysis Date31/01/2025 Catalog NumberK182001
Lot Number2425241 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog NumberK182001
Lot Number2463685 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog NumberK182001
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Invitrogen™ PureLink™ Genomic DNA Mini Kit > 50 Preps

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.