Learn More
Pepsin, Extra Pure, Powder, SLR, Fisher Chemical™
Enzyme that breaks down proteins into smaller peptides.
Brand: Fisher Chemical P/1120/46
35.69 GBP valid until 2025-04-30
Use promo code "24155" to get your promotional price.
Description
Enzyme that breaks down proteins into smaller peptides.
Proteolitic activity BPC: 2500 - 3000 (Based on the British Pharmaceutical Codex 1959 (BPC) for the pepsin proteolytic activity report. Dissolves not less than 2500 times its weight of coagulated egg albumin.
Specifications
25 g | |
Pepsin | |
Powder |
Product Suggestions
Customers who viewed this item also viewed.
Safety and Handling
- Pepsin
Signal Word
- Danger
Hazard Category
- Skin corrosion/irritation Category 2
- Serious eye damage/eye irritation Category 2
- Respiratory sensitiser Category 1
- Specific target organ toxicity Category 3
Hazard Statement
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
- H335-May cause respiratory irritation.
Precautionary Statement
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.