missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cole-Parmer™ Peroxide-Cured Silicone Tubing
Flexible tubing offers long shelf life and resists kinking and tearing.
Brand: Cole-Parmer™ 50121-44
This item is not returnable.
View return policy
Description
- Exhibits good heat, cold, ozone, UV, and abrasion resistance
- Ideal for general-purpose, laboratory, and single-use applications
- Free technical application support available to assist with tubing selection
Specifications
RoHS, ADCF | |
3/16 in. | |
Rubber | |
8 mm | |
7.62 m | |
1.6 mm | |
-56°C | |
Very Flexible | |
204°C |
Not Autoclavable | |
4.8 mm | |
5/16 in. | |
25 ft. | |
1/16 in. | |
-70°F | |
Clear | |
400°F | |
Tubing |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-5 of 11
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Cole-Parmer™ Peroxide-Cured Silicone Tubing > Tubing ID: 3/16 in.; Tubing OD: 5/16 in.; Wall Thickness: 1/16 in.
This product is not approved or intended for, and should not be used for, medical, clinical, surgical or other patient-oriented applications.
Spot an opportunity for improvement?Share a Content Correction